Learn More
Aviva Systems Biology Corp SEMG1 Antibody - N-terminal region (ARP42088_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42088_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMG1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SEMG1. Alternate names: MGC14719, SEMG, SGI, CT103, dJ172H20.2ApplicationIHC, WB. Species reactivity: Human, Pig
Catalog No. 50-014-58102