Learn More
Aviva Systems Biology Corp SEMG1 Antibody - middle region (ARP42089_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42089_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEMG1. Alternate names: MGC14719, SEMG, SGI, CT103, dJ172H20.2ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-58103