Learn More
Aviva Systems Biology Corp SEMG2 Antibody - N-terminal region (ARP42092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SEMG2. Alternate names: SGIIApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-284-976