Learn More
Yamato Scientific America, Inc. Yamato Semi-Automatic Benchtop Glassware Laboratory Washer, 115V/220V.
Catalog No. 501898202 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Semi automatic benchtop glassware washer with upward and downward two-way pressurized water with rotating jet nozzles bringing high level cleaning. Easy to to operate by simply setting time then start. With built-in water heater, no boiler piping and water heating system are required. Optional jet rack is available for hard to clean targets, such as glassware with narrow neck or body. Washing water temp. range: ambient ~60°C.
Catalog No. 50-189-8202 Supplier Yamato Scientific America, Inc. Supplier No. AW47115V / AW47220V
Add to Cart
Add to Cart
Request a Quote
Semi automatic benchtop glassware washer with upward and downward two-way pressurized water with rotating jet nozzles bringing high level cleaning. Easy to to operate by simply setting time then start. With built-in water heater, no boiler piping and water heating system are required. Optional jet rack is available for hard to clean targets, such as glassware with narrow neck or body. Washing water temp. range: ambient ~60°C.