Learn More
Glass Expansion Semi Demountable Quartz Torch Body, 6mm OD Side Arms for Finnigan Element
SDP
Catalog No. 502703161
Encompass_Preferred
Semi Demountable Quartz Torch Body, 6mm OD Side Arms for Thermo (Finnigan) Element 1, Element 2, Neptune ICP-MS. Requires Glass Expansion’s Semi Demountable Torch Adaptor 31-808-0459. Injector options include Tapered Quartz Injector 1.5mm (501907057, 31-808-0007), and Tapered Alumina Injector 1.8mm (501907063, 31-808-0015). GazFit Connectors for 6mm OD Side Arm (GAZ-06), and the Extractor Tool for Semi Demountable Torch, 12.1mm, Finnigan (31-100-0109).Assembled Semi Demountable Torches are available with 1.5mm Tapered Quartz Injector (31-808-0394) and with 1.8mm Tapered Alumina Injector (31-808-0395).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3161 Supplier Glass Expansion Supplier No. 318080451
Semi Demountable Quartz Torch Body, 6mm OD Side Arms for Thermo (Finnigan) Element 1, Element 2, Neptune ICP-MS. Requires Glass Expansion’s Semi Demountable Torch Adaptor 31-808-0459. Injector options include Tapered Quartz Injector 1.5mm (501907057, 31-808-0007), and Tapered Alumina Injector 1.8mm (501907063, 31-808-0015). GazFit Connectors for 6mm OD Side Arm (GAZ-06), and the Extractor Tool for Semi Demountable Torch, 12.1mm, Finnigan (31-100-0109).Assembled Semi Demountable Torches are available with 1.5mm Tapered Quartz Injector (31-808-0394) and with 1.8mm Tapered Alumina Injector (31-808-0395).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001