Learn More
Glass Expansion Semi-Demountable Torch with 2.4mm Alumina Injector Agilent 7700/7800/7900/8800/8900 ICP-MS Injector options: Sapphire 2.4mm Capillary Injector 31-808-4159 and 2.4mm Tapered Quartz Injector 31-808-4168
SDP
Catalog No. 502703163
Encompass_Preferred
Semi Demountable Torch (SDT) with 2.4mm Alumina Injector. Suitable for Agilent Technologies 7700/7800/7900/8800 and 8900 ICP-MS.This Semi-demountable quartz torch with alumina injector is suitable for use with aqueous, including HF, and organic solvent-based matrices. Optional injectors include 2.4 mm Tapered Quartz Injector (NC2046978) and 2.4 mm Tapered Sapphire Injector (NC2046976). Replacement parts include the Injector Adaptor (31-808-4129) and Semi Demountable Torch Body (NC2046979).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3163 Supplier Glass Expansion Supplier No. 308084185
Semi Demountable Torch (SDT) with 2.4mm Alumina Injector. Suitable for Agilent Technologies 7700/7800/7900/8800 and 8900 ICP-MS.This Semi-demountable quartz torch with alumina injector is suitable for use with aqueous, including HF, and organic solvent-based matrices. Optional injectors include 2.4 mm Tapered Quartz Injector (NC2046978) and 2.4 mm Tapered Sapphire Injector (NC2046976). Replacement parts include the Injector Adaptor (31-808-4129) and Semi Demountable Torch Body (NC2046979).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001