Learn More
Glass Expansion Semi-Demountable Torch with 1.5mm Alumina Injector for Shimadzu
SDP
Catalog No. 502703162
Encompass_Preferred
Semi-Demountable Torch with 1.5mm Alumina Injector (replaces 30-808-8039). Compatible with Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES. Includes Semi Demountable Torch Body 31-808-4341, Injector Adaptor for Semi Demountable Torch 31-808-4129, and Tapered Alumina Injector 31-808-3202.Replacement Viton O-Rings (PKT 10) are 70-V-011.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3162 Supplier Glass Expansion Supplier No. 308084335
Semi-Demountable Torch with 1.5mm Alumina Injector (replaces 30-808-8039). Compatible with Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES. Includes Semi Demountable Torch Body 31-808-4341, Injector Adaptor for Semi Demountable Torch 31-808-4129, and Tapered Alumina Injector 31-808-3202.Replacement Viton O-Rings (PKT 10) are 70-V-011.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001