Learn More
Glass Expansion Semi-Demountable Torch Adaptor Thermo X-Series
SDP
Catalog No. 502703158
Encompass_Preferred
Semi-Demountable Torch Adaptor. Compatible with Glass Expansion’s semi-demountable torch for Thermo X-Series ICP-MS. For use with X-Series semi-demountable torch body 31-808-1009. Injector options include Tapered Alumina Injector 1.5mm (501907125, 31-808-1013), Sapphire Injector 2.0mm (501907171, 31-808-3001) and Platinum Injector 1.5mm (31-808-1256).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3158 Supplier Glass Expansion Supplier No. 318080984
Semi-Demountable Torch Adaptor. Compatible with Glass Expansion’s semi-demountable torch for Thermo X-Series ICP-MS. For use with X-Series semi-demountable torch body 31-808-1009. Injector options include Tapered Alumina Injector 1.5mm (501907125, 31-808-1013), Sapphire Injector 2.0mm (501907171, 31-808-3001) and Platinum Injector 1.5mm (31-808-1256).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001