Learn More
Glass Expansion Semi-Demountable Torch Adaptor Thermo (Finnigan) Element 1, Element 2, Neptune
SDP
Catalog No. 502703160
Encompass_Preferred
Semi-Demountable Torch Adaptor. Compatible with Glass Expansion’s Semi Demountable Torch Body (31-808-1009) for Thermo (Finnigan) Element 1, Element 2, Neptune.Replacement O-Ring Kit is 70-0459.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3160 Supplier Glass Expansion Supplier No. 318080459
Semi-Demountable Torch Adaptor. Compatible with Glass Expansion’s Semi Demountable Torch Body (31-808-1009) for Thermo (Finnigan) Element 1, Element 2, Neptune.Replacement O-Ring Kit is 70-0459.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001