Learn More
Glass Expansion Semi Demountable Torch Body
SDP
Catalog No. 502703098
Encompass_Preferred
Semi Demountable Torch Body. Compatible with Glass Expansion’s Semi Demountable Torch for Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES (30-808-4335).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3098 Supplier Glass Expansion Supplier No. 318084341
Semi Demountable Torch Body. Compatible with Glass Expansion’s Semi Demountable Torch for Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES (30-808-4335).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001