Learn More
Glass Expansion Semi-Demountable Torch Body Thermo X-Series
SDP
Catalog No. 502703157
Encompass_Preferred
Semi-Demountable Torch Body. Compatible with Thermo X-Series ICP-MS. For use with Glass Expansion’s semi-demountable torch adaptor 31-808-0984. Injector options include Tapered Alumina Injector 1.5mm (501907125, 31-808-1013), Sapphire Injector 2.0mm (501907171, 31-808-3001) and Platinum Injector 1.5mm (31-808-1256).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3157 Supplier Glass Expansion Supplier No. 318081009
Semi-Demountable Torch Body. Compatible with Thermo X-Series ICP-MS. For use with Glass Expansion’s semi-demountable torch adaptor 31-808-0984. Injector options include Tapered Alumina Injector 1.5mm (501907125, 31-808-1013), Sapphire Injector 2.0mm (501907171, 31-808-3001) and Platinum Injector 1.5mm (31-808-1256).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001