Learn More
Glass Expansion Semi-Demountable Torch with 2.4mm Quartz Injector for 7700, 7800, 7900, 8800 & 8900 ICP-MS
SDP
Catalog No. NC2046974
Encompass_Preferred
Semi Demountable Torch (SDT) with 2.4mm Quartz Injector. Suitable for Agilent Technologies 7700/7800/7900/8800 and 8900 ICP-MS.Semi-demountable quartz torch suitable for routine aqueous and high dissolved solids applications but not inert to samples containing hydrofluoric acid (HF). Optional injectors include 2.4 mm Tapered Alumina Injector (50-190-7264) and 2.4 mm Tapered Sapphire Injector (NC2046976). Replacement parts include the Injector Adaptor (31-808-4129) and Semi Demountable Torch Body (NC2046979).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2046974 Supplier Glass Expansion Supplier No. 308083853
Semi Demountable Torch (SDT) with 2.4mm Quartz Injector. Suitable for Agilent Technologies 7700/7800/7900/8800 and 8900 ICP-MS.Semi-demountable quartz torch suitable for routine aqueous and high dissolved solids applications but not inert to samples containing hydrofluoric acid (HF). Optional injectors include 2.4 mm Tapered Alumina Injector (50-190-7264) and 2.4 mm Tapered Sapphire Injector (NC2046976). Replacement parts include the Injector Adaptor (31-808-4129) and Semi Demountable Torch Body (NC2046979).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001