Learn More
Azzota Corp Semi-Micro 10mm blk 1.4ml Qz
Catalog No. 5000887135
Encompass_Preferred
Semi-Micro cell with black walls and lid 10mm Pathlength 4mm Inside Width Volume - 1 4ml Quartz
Catalog No. 50-008-87135 Supplier Azzota Corp Supplier No. Q24
Semi-Micro cell with black walls and lid 10mm Pathlength 4mm Inside Width Volume - 1 4ml Quartz