Learn More
Azzota Corp Semi-Micro 30mm blk 4.2ml Qz
Catalog No. 5000887063
Encompass_Preferred
Semi-Micro cell with black walls and lid 30mm Pathlength 4mm Inside Width Volume - 4 2ml Quartz
Catalog No. 50-008-87063 Supplier Azzota Corp Supplier No. Q26
Semi-Micro cell with black walls and lid 30mm Pathlength 4mm Inside Width Volume - 4 2ml Quartz