Learn More
Azzota Corp Semi-Micro 5mm blk 0.7ml Qz
Catalog No. 5000887134
Encompass_Preferred
Semi-Micro cell with black walls and lid 5mm Pathlength 4mm Inside Width Volume - 0 7ml Quartz
Catalog No. 50-008-87134 Supplier Azzota Corp Supplier No. Q23
Semi-Micro cell with black walls and lid 5mm Pathlength 4mm Inside Width Volume - 0 7ml Quartz