Learn More
Starna Cells Inc SEMI MICRO CELL
SDP
Catalog No. NC9884429
Encompass_Preferred
Sub-Micro Fluorometer Cells, Type 16F, Window material: Spectrosil Quartz, Useable Range: 170 to 2700nm, Path Length: 10mm, Aqueous Volume: 65 microliter, Exterior: 12.5L x 12.5W x 45mmH, Window: 2W x 2.5mmH, Z Dimension: 8.5mm, Geometric V
Catalog No. NC9884429 Supplier Starna Cells Inc Supplier No. 16.50Q10/Z8.5
Sub-Micro Fluorometer Cells, Type 16F, Window material: Spectrosil Quartz, Useable Range: 170 to 2700nm, Path Length: 10mm, Aqueous Volume: 65 microliter, Exterior: 12.5L x 12.5W x 45mmH, Window: 2W x 2.5mmH, Z Dimension: 8.5mm, Geometric V