Learn More
World Precision Instrument Semi Micro Concentric Bipolar Electrodes with Extension, Rhodes Style Tip-SSM04RC100
SDP
Catalog No. 502538564
Encompass
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.
Catalog No. 50-253-8564 Supplier World Precision Instrument Supplier No. SSM04RC100
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.