Learn More
World Precision Instrument Semi Micro Concentric Bipolar Electrodes, Rhodes Style Tip-PTM321RC100
SDP
Catalog No. 502538623
Encompass
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.
Catalog No. 50-253-8623 Supplier World Precision Instrument Supplier No. PTM321RC100
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.