Learn More
World Precision Instrument Semi Micro Concentric Bipolar Electrodes with Extension, Rhodes Style Tip-PTM04RC100
SDP
Catalog No. 502538627
Encompass
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.
Catalog No. 50-253-8627 Supplier World Precision Instrument Supplier No. PTM04RC100
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.