Learn More
Starna Cells Inc SEMI MICRO FLUOROMETER QUARTZ
SDP
Catalog No. NC9695773
Encompass_Preferred
Semi-Micro Fluorometer Cell, Open Top, Type: 9F, Window Material: Spectrosil quartz, Useable Range: 170 to 2700nm, Path Length: 10mm, Exterior: 12.5L x 12.5H x 45mmW, Interior: 4W x 10mm long, Nominal Volume: 1.400m long.
Catalog No. NC9695773 Supplier Starna Cells Inc Supplier No. 9FQ10
Semi-Micro Fluorometer Cell, Open Top, Type: 9F, Window Material: Spectrosil quartz, Useable Range: 170 to 2700nm, Path Length: 10mm, Exterior: 12.5L x 12.5H x 45mmW, Interior: 4W x 10mm long, Nominal Volume: 1.400m long.