Learn More
Andwin Scientific SEMI MICRO PS CUVETTES 500/CS
SDP
Catalog No. NC9626091
Encompass_Preferred
The Plastic Cuvettes are disposable. This product has a volume capacity of 1.5 mL. It is sold in a case of 500.
Catalog No. NC9626091 Supplier Andwin Scientific Supplier No. 58017847
The Plastic Cuvettes are disposable. This product has a volume capacity of 1.5 mL. It is sold in a case of 500.