Learn More
Andwin Scientific SEMI MICRO PS CUVETTES 500/CS
SDP
Supplier:  Andwin Scientific 97000586
Encompass_Preferred
The two-sided disposable semi-micro plastic cuvette can be used with most spectrophotometers and photometers. This is a superior product for most spectrophotometry applications in the visible and near-UV range. The cuvettes feature a 10 mm light path and include an arrow mark that can display the direction of transmission and reduce variation. This product has a volume range of 1.53.0 mL and is sold in a case of 500.
Catalog No. NC9697862