Learn More
Starna Cells Inc SEMI MICRO QUARTZ
SDP
Catalog No. NC9082994
Encompass_Preferred
Starna Semi-Micro Cells, Window material Spectrosil Quartz or equivalent, Usable range: 170 to 2700nm, Path Length: 10mm, External W and L and H 12.5mm and 12.5mm and 45mm, Internal W and L, 4mm and 10mm, Nominal Volume: 1.160mL
Catalog No. NC9082994 Supplier Starna Cells Inc Supplier No. 9/9Q10
Starna Semi-Micro Cells, Window material Spectrosil Quartz or equivalent, Usable range: 170 to 2700nm, Path Length: 10mm, External W and L and H 12.5mm and 12.5mm and 45mm, Internal W and L, 4mm and 10mm, Nominal Volume: 1.160mL