Learn More
Starna Cells Inc SEMI MICRO QUARTZ CUVETTE 1/EA
SDP
Catalog No. NC9871235
Encompass_Preferred
Semi Micro Quartz Cuvette, Path Length: 10mm, Exterior Dimension: 12.5L x 12.5W x 45mmH, Interior Dimension: 10L x 4W, Nominal Volume: 1.400 mL, Window Material: Spectrosil UV Quartz windows, Useable Range: 170 to 2700nm, Polished Windows:
Catalog No. NC9871235 Supplier Starna Cells Inc Supplier No. 9BQ10
Semi Micro Quartz Cuvette, Path Length: 10mm, Exterior Dimension: 12.5L x 12.5W x 45mmH, Interior Dimension: 10L x 4W, Nominal Volume: 1.400 mL, Window Material: Spectrosil UV Quartz windows, Useable Range: 170 to 2700nm, Polished Windows: