Learn More
Starna Cells Inc SEMI MICRO SELF MSKG QTZ CELL
SDP
Catalog No. NC9276411
Encompass_Preferred
Micro Spectrophotometer Cells, Window Material: Spectrosil; Quartz, Useable Range: 170 to 2700nm, Path Length: 10mm, Exterior Size: 12.5W x 12.5 L x 45mmH, Interior Size: 4W x 10mm L, Base Thickness: 9mm, Nominal Volume: 1.160mL
Catalog No. NC9276411 Supplier Starna Cells Inc Supplier No. 9B9Q10
Micro Spectrophotometer Cells, Window Material: Spectrosil; Quartz, Useable Range: 170 to 2700nm, Path Length: 10mm, Exterior Size: 12.5W x 12.5 L x 45mmH, Interior Size: 4W x 10mm L, Base Thickness: 9mm, Nominal Volume: 1.160mL