Learn More
Analytical Sales & Services Semi-Prep Adjustable Flow Splitter, Post Column
Catalog No. 5000806144
Encompass
Advantage Model 62 Semi Prep Adjustable Flow Splitter 1 000:1 to 20 000:1 ratio Post Column
Qty/Pkg: EACH
Catalog No. 50-008-06144 Supplier Analytical Sales & Services Supplier No. 62311
Advantage Model 62 Semi Prep Adjustable Flow Splitter 1 000:1 to 20 000:1 ratio Post Column
Qty/Pkg: EACH