Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, Diamond Hydride, 4um, 100A. 10mm ID x 250mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 70000SP250
Encompass_Preferred
This Cogent Diamond Hydride "Semi" Preparative, HPLC Column is made with 4um, 100A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for Purification or "Prep" of Peptides, Metabolites, Polar Compounds and other Analytes with amine groups when a large volume of sample is desired to be collected. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0947