Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, HPS Amino, 5um, 120A. 10mm ID x 250mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 75040SP250
Encompass_Preferred
This Cogent HPS Amino™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. Not end capped, it is manufactured using tri-functional bonding of a 3 carbon silane with a terminal amino group to our unique High Purity Silica. This Column is ideal for new, small volume prep methods. With a 5 µm Particle, this Column works well with higher flow rates and lower back pressures compared with smaller particles which is desirable for Semi Preparative Chromatography. Used for polar molecules with a negative charge, such compounds with Carboxylic Acid Groups and Sugars, in Normal Phase or HILIC modes. Compares to Inertsil™ Amino and Luna™ Amino Columns in performance. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-1051