Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, RP C18 Wide Pore, 10um, 300A. 10mm ID x 100mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68018SP1003M
Encompass_Preferred
This Cogent RP C18™, 300Å, Wide Pore Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. This makes this column ideal as an Alternate in USP Methods that use the L1 Classification, for large molecules.With a 10 µm Particle and 300A Pore Size, this column works well with higher flow rates and lower back pressures. The diameter and length of this column provides increased loading capacity compared with analytical columns, which is why it is chosen for purification of large hydrophobic or neutral compounds, making it ideal for Semi Preparative HPLC. A specialized HPLC System is required for columns this size.
Catalog No. 50-226-9829