Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, RP C18 Wide Pore, 5um, 300A. 10mm ID x 250mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68518SP2503M
Encompass_Preferred
This Cogent RP C18™, 300Å, Wide Pore Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. With a 5µm Particle and 300A Pore Size, this column works well with higher flow rates and lower back pressures. The diameter and long length of this column provides increased loading capacity compared with analytical columns, which is why it is chosen for purification of large hydrophobic or neutral compounds, making it ideal for Semi Preparative HPLC. A specialized HPLC System is required for columns this size.
Catalog No. 50-226-9891