Learn More
Analytical Sales & Services Semi-Prep Guard Column Holder
Catalog No. 5000804921
Encompass
Semi-Prep Guard Column Holder. Holds 10mm x 1cm guard cartridge, 780µL internal volume
Catalog No. 50-008-04921 Supplier Analytical Sales & Services Supplier No. 521000
Semi-Prep Guard Column Holder. Holds 10mm x 1cm guard cartridge, 780µL internal volume