Learn More
Cayman Chemical SemI-synthetIc FerutInIn 10mg
SDP
Catalog No. 502968430
Encompass_Preferred
A plant-derived ester of a sesquiterpenic alcohol that acts as an agonist for ERα (IC50 = 33.1 nM) and an agonist/antagonist for ERβ (IC50 = 180.5 nM); demonstrates ionophorectic properties, increasing calcium permeability of lipid bilayer membranes, mitochondria, thymocytes, sarcoplasmic reticulum, and liposomes at 1-50 µM
Catalog No. 50-296-8430 Supplier Cayman Chemical Supplier No. 1655410 MG
A plant-derived ester of a sesquiterpenic alcohol that acts as an agonist for ERα (IC50 = 33.1 nM) and an agonist/antagonist for ERβ (IC50 = 180.5 nM); demonstrates ionophorectic properties, increasing calcium permeability of lipid bilayer membranes, mitochondria, thymocytes, sarcoplasmic reticulum, and liposomes at 1-50 µM