Learn More
Supelco Inc SEMI-VOLATILE CAL. MIX STD 1ML
Catalog No. NC9653592
Encompass
CLP Semivolatile Calibration Mix 1mL Solvent: methylene chloride:benzene (3:1) Hazard Symbol: F TStorage Temp: FREEZE
Catalog No. NC9653592 Supplier Supelco Inc Supplier No. 506508
CLP Semivolatile Calibration Mix 1mL Solvent: methylene chloride:benzene (3:1) Hazard Symbol: F TStorage Temp: FREEZE