Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semi-Volatile Calibration Standard, SPEX CertiPrep™
SDP

Catalog No. BIGBN25 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BIG-BN-2-5 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BIG-BN-2-5 Supplier SPEX CertiPrep Supplier No. BIGBN25

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2,4-Trichlorobenzene120-82-12000μg/mL
1,2-Dichlorobenzene95-50-12000μg/mL
1,3-Dichlorobenzene541-73-12000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
2,4-Dinitrotoluene121-14-22000μg/mL
2,6-Dinitrotoluene606-20-22000μg/mL
2-Chloronaphthalene91-58-72000μg/mL
2-Methylnaphthalene91-57-62000μg/mL
2-Nitroaniline88-74-42000μg/mL
3-Nitroaniline99-09-22000μg/mL
4-Bromodiphenyl ether101-55-32000μg/mL
4-Chloroaniline (p-Chloroaniline)106-47-82000μg/mL
4-Chlorophenyl-phenyl ether7005-72-32000μg/mL
4-Nitroaniline100-01-62000μg/mL
Acenaphthene83-32-92000μg/mL
Acenaphthylene208-96-82000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(g,h,i)perylene191-24-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Benzyl alcohol100-51-62000μg/mL
Bis(2-chloro-1-methylethyl) ether108-60-12000μg/mL
bis(2-Chloroethoxy)methane111-91-12000μg/mL
bis(2-Chloroethyl)ether111-44-42000μg/mL
Bis(2-Ethylhexyl)phthalate117-81-72000μg/mL
Butylbenzyl phthalate85-68-72000μg/mL
Chrysene218-01-92000μg/mL
Dibenz(a,h)anthracene53-70-32000μg/mL
Dibenzofuran132-64-92000μg/mL
Diethyl phthalate84-66-22000μg/mL
Dimethyl phthalate131-11-32000μg/mL
Di-n-butyl phthalate84-74-22000μg/mL
Di-n-octyl phthalate117-84-02000μg/mL
Fluoranthene206-44-02000μg/mL
Fluorene86-73-72000μg/mL
Hexachlorobenzene118-74-12000μg/mL
Hexachlorobutadiene87-68-32000μg/mL
Hexachlorocyclopentadiene77-47-42000μg/mL
Hexachloroethane67-72-12000μg/mL
Indeno(1,2,3-c,d)pyrene193-39-52000μg/mL
Isophorone78-59-12000μg/mL
Naphthalene91-20-32000μg/mL
Nitrobenzene98-95-32000μg/mL
N-Nitrosodi-n-propylamine621-64-72000μg/mL
N-Nitrosodiphenylamine86-30-62000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Semi-volatile Calibration Standard
Packaging Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Methods 625, 8270, CLP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methylene Chloride/Benzene
Quantity 5 mL
Concentration or Composition (by Analyte or Components) Components in Methylene Chloride:Benzene at 2000μg/mL
Acenaphthene    Dimethyl Phthalate
Acenaphthylene    Di-n-butyl Phthalate
Anthracene    2,4-Dinitrotoluene
Benzo(a)anthracene    2,6-Dinitrotoluene
Benzo(a)pyrene    Di-n-octyl Phthalate
Benzo(b)fluoranthene    Fluoranthene
Benzo(g,h,i)perylene    Fluorene
Benzo(k)fluoranthene    Hexachlorobenzene
Benzyl Alcohol    Hexachlorobutadiene
bis(2-chloroethoxy)methane    Hexachlorocyclopentadiene
bis(2-chloroethyl)ether    Hexachloroethane
bis(2-chloroisopropyl)ether    Indeno(1,2,3-cd)pyrene
bis(2-ethylhexyl)phthalate    Isophorone
4-Bromophenyl phenyl ether    2-Methylnaphthalene
Butylbenzyl phthalate    Naphthalene
4-chloroaniline    2-Nitroaniline
2-chloronaphthalene    3-Nitroaniline
4-chlorophenyl-phenyl ether    4-Nitroaniline
Chrysene    Nitrobenzene
Dibenz(a,h)anthracene    N-Nitroso-di-n-propylamine
Dibenzofuran    N-Nitrosodiphenylamine
1,2-Dichlorobenzene    Phenanthrene
1,3-Dichlorobenzene    Pyrene
1,4-Dichlorobenzene    1,2,4-Trichlorobenzene
Diethyl Phthalate

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.