Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semi-Volatiles Internal Standard Mixture (For EPA 625, 8270C, CLP)

Catalog No. usus108n
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-US-108N 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-US-108N Supplier Agilent Technologies Supplier No. US108N1
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Semi-Volatiles Internal Standard Mixture
Packaging Ampule
Molecular Formula CH2Cl2
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 4000μg/mL in Methylene Chloride:
Acenaphthene-d10
Chrysene-d12
1,4-Dichlorobenzene-d4
Naphthalene-d8
Perylene-d12
Phenanthrene-d10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.