Learn More
Semi-Volatiles Mixture No. 3, Quantitative, 18-Analyte, 1 mL
Specifications
Specifications
| Chemical Name or Material | Semi-Volatiles Mixture No. 3 |
| CAS Max % | 97.28, 0.15, 0.15, 0.15, 0.15, 0.15 |
| CAS | 98-86-2, 65-85-0, 111-91-1, 106-47-8, 59-50-7 |
| Analyte or Component Names | methylene chloride (dichloromethane),acetophenone,benzoic acid |
| Element | Methylene Chloride, Acetophenone, Benzoic Acid |
| Solvent or Matrix | Methylene Chloride |
| Quantity | 1 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 2000μg/mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.