Learn More
Medchemexpress LLC Semicarbazide-13C,15N2 hydrochloride | 1173020-16-0 | MFCD06201846 | 99.9% | 114.51 g/mol | CH6ClN3O | 10 MG
SDP
Catalog No. 5000210740
Encompass_Preferred
Semicarbazide-13C,15N2 hydrochloride is an isotopically labeled semicarbazide hydrochloride supplied as a solid reference standard for analytical and tracer applications.
  • Isotopically labeled with 13C and two 15N atoms.
  • High purity suitable for analytical work (99.9%).
  • Provided as a white to off-white solid.
  • Intended for use as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Store sealed at -20°C; in solution, store at -80°C up to 6 months.
Catalog No. 50-002-10740 Supplier Medchemexpress LLC Supplier No. HYW067028S10MG
Semicarbazide-13C,15N2 hydrochloride is an isotopically labeled semicarbazide hydrochloride supplied as a solid reference standard for analytical and tracer applications.
  • Isotopically labeled with 13C and two 15N atoms.
  • High purity suitable for analytical work (99.9%).
  • Provided as a white to off-white solid.
  • Intended for use as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Store sealed at -20°C; in solution, store at -80°C up to 6 months.