Learn More
Medchemexpress LLC Semicarbazide (hydrochloride) | 563-41-7 | MFCD00013009 | 98.0% | 111.53 | CH6ClN3O | 50g
SDP
Catalog No. 5000269965
Encompass_Preferred
Semicarbazide hydrochloride is an orally active urea derivative Semicarbazide hydrochloride binds to copper or iron in cells Semicarbazide hydrochloride inhibits the activity of soluble semicarbazide sensitive amine oxidase (SSAO) Semicarbazide hydrochloride damages cartilage blood vessels ovaries testicles and thyroid follicles[1][2][3][4][5][6][7][8][9]
Catalog No. 50-002-69965 Supplier Medchemexpress LLC Supplier No. HYY047050G
Semicarbazide hydrochloride is an orally active urea derivative Semicarbazide hydrochloride binds to copper or iron in cells Semicarbazide hydrochloride inhibits the activity of soluble semicarbazide sensitive amine oxidase (SSAO) Semicarbazide hydrochloride damages cartilage blood vessels ovaries testicles and thyroid follicles[1][2][3][4][5][6][7][8][9]