Learn More
Medchemexpress LLC Semicarbazide hydrochloride | 563-41-7 | MFCD00013009 | 98.0% | 111.53 g/mol | CH6ClN3O | 1mL
SDP
Catalog No. 5000269964
Encompass_Preferred
Semicarbazide hydrochloride is an orally active urea derivative Semicarbazide hydrochloride binds to copper or iron in cells Semicarbazide hydrochloride inhibits the activity of soluble semicarbazide sensitive amine oxidase (SSAO) Semicarbazide hydrochloride damages cartilage blood vessels ovaries testicles and thyroid follicles[1][2][3][4][5][6][7][8][9]
Catalog No. 50-002-69964 Supplier Medchemexpress LLC Supplier No. HYY047010MM/1ML
Add to Cart
Add to Cart
Semicarbazide hydrochloride is an orally active urea derivative Semicarbazide hydrochloride binds to copper or iron in cells Semicarbazide hydrochloride inhibits the activity of soluble semicarbazide sensitive amine oxidase (SSAO) Semicarbazide hydrochloride damages cartilage blood vessels ovaries testicles and thyroid follicles[1][2][3][4][5][6][7][8][9]