Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semicarbazide Hydrochloride, 99%, (Crystalline), MP Biomedicals

Catalog No. ICN10286480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10286480 100 g
ICN10286490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10286480 Supplier MP Biomedicals, Inc Supplier No. 0210286480

Urease substrate and MAO inhibitor

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-41-7
Molecular Formula CH6ClN3O
Molecular Weight (g/mol) 111.53
MDL Number MFCD00013009
InChI Key XHQYBDSXTDXSHY-UHFFFAOYSA-N
Synonym semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
PubChem CID 11236
ChEBI CHEBI:82532
IUPAC Name hydrogen aminourea chloride
SMILES [H+].[Cl-].NNC(N)=O

Specifications

Assay Percent Range 99%
Quantity 100 g
Formula Weight 111.5
Physical Form Crystalline
Chemical Name or Material Semicarbazide Hydrochloride
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.