Learn More
Grainger SemiCondctveAbrsnRstntTape 1inx18yd 7mil
Catalog No. 502749772
Encompass_Preferred
The TapeCase Insulating Electrical Tape is a high-quality electrical tape made in the USA by TapeCase. It is a 7 mil thick PVC insulating tape with an operating temperature range of up to 105°C. The tape has an Underwriters Laboratories listed flame retardant rating and is designed for general purpose electrical insulation, harnessing, bundling, sealing and moisture proofing applications. It comes in a variety of colors and is sold in multiple widths and lengths by the roll. The tape provides excellent mechanical and electrical protection.
Catalog No. 50-274-9772 Supplier Grainger Supplier No. 15D540
The TapeCase Insulating Electrical Tape is a high-quality electrical tape made in the USA by TapeCase. It is a 7 mil thick PVC insulating tape with an operating temperature range of up to 105°C. The tape has an Underwriters Laboratories listed flame retardant rating and is designed for general purpose electrical insulation, harnessing, bundling, sealing and moisture proofing applications. It comes in a variety of colors and is sold in multiple widths and lengths by the roll. The tape provides excellent mechanical and electrical protection.