Learn More
Grainger SemiCondctveAbrsnRstntTape 2inx18yd 7mil
Catalog No. 502749402
Encompass_Preferred
The TapeCase Insulating Electrical Tape is a high-quality electrical tape made in the USA by TapeCase. It is a 7 mil thick PVC insulating tape with an operating temperature range of up to 105°C. The tape has an Underwriters Laboratories UL510 flame retardant rating and is designed for insulating and sealing electrical wires and cables. This 3/4" x 60' roll tape offers high dielectric strength, flexibility, and adhesion. It can be used for general purpose electrical insulation, harnessing, banding, sealing, and more.
Catalog No. 50-274-9402 Supplier Grainger Supplier No. 15D541
The TapeCase Insulating Electrical Tape is a high-quality electrical tape made in the USA by TapeCase. It is a 7 mil thick PVC insulating tape with an operating temperature range of up to 105°C. The tape has an Underwriters Laboratories UL510 flame retardant rating and is designed for insulating and sealing electrical wires and cables. This 3/4" x 60' roll tape offers high dielectric strength, flexibility, and adhesion. It can be used for general purpose electrical insulation, harnessing, banding, sealing, and more.