Learn More
Grainger SemiCondctveAbrsnRstntTape 3inx18yd 7mil
Catalog No. 502751826
Encompass_Preferred
The TAPECASE Cloth Tape PTFE Slick Surface is a high-quality cloth tape with a PTFE-coated surface that provides a slick, non-stick finish. This general purpose tape has a tensile strength of 40 lbs per inch width and adheres to a variety of surfaces including glass, plastic, rubber, wood, and metal. It remains flexible from -20°F to 250°F. This 3/4" x 60 yard roll of tan cloth tape is made in the USA by TapeCase, a leading manufacturer of specialty tapes, adhesives, and films. The slick PTFE surface resists adhesives and leaves no residue behind.
Catalog No. 50-275-1826 Supplier Grainger Supplier No. 15D542
The TAPECASE Cloth Tape PTFE Slick Surface is a high-quality cloth tape with a PTFE-coated surface that provides a slick, non-stick finish. This general purpose tape has a tensile strength of 40 lbs per inch width and adheres to a variety of surfaces including glass, plastic, rubber, wood, and metal. It remains flexible from -20°F to 250°F. This 3/4" x 60 yard roll of tan cloth tape is made in the USA by TapeCase, a leading manufacturer of specialty tapes, adhesives, and films. The slick PTFE surface resists adhesives and leaves no residue behind.