Learn More
Starna Cells Inc SEMIMICR SQ CELL9 10MM Q 1.4ML
SDP
Catalog No. NC9494012
Encompass_Preferred
Spectrosil Far UV Quartz windows Useable range: 170 to 2700nm Pathlength: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width: 4mm Interior Length: 10mm Nominal Volume: 1.4mL Number polished windows:2
Catalog No. NC9494012 Supplier Starna Cells Inc Supplier No. 9Q10
Spectrosil Far UV Quartz windows Useable range: 170 to 2700nm Pathlength: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width: 4mm Interior Length: 10mm Nominal Volume: 1.4mL Number polished windows:2