Learn More
Starna Cells Inc SEMIMICRO FLORO QUARTZ CELL
SDP
Catalog No. NC9785751
Encompass_Preferred
Semi-Micro Fluorometer Cell, open top, Window material: Spectrosil Quartz, useable range:170 to 2700 nm, Path Length:10 mm, Exterior (WxLxH): 12.5 x 12.5 x 45 mm, Interior (W x L): 4 x 10 mm, Nominal Volume :1.400 ml, Number Polished Window
Catalog No. NC9785751 Supplier Starna Cells Inc Supplier No. 9FQ10GL14S
Semi-Micro Fluorometer Cell, open top, Window material: Spectrosil Quartz, useable range:170 to 2700 nm, Path Length:10 mm, Exterior (WxLxH): 12.5 x 12.5 x 45 mm, Interior (W x L): 4 x 10 mm, Nominal Volume :1.400 ml, Number Polished Window