Learn More
WELCH MATERIALS INC Direct Connect Semi-prep guard column holder(10mm)
Catalog No. NC3779987
Encompass_Preferred
Direct Connect Semi-prep guard column holder(10mm) × 1, for 10×10mm Semi-Prep Cartridge
Catalog No. NC3779987 Supplier WELCH MATERIALS INC Supplier No. 0080801116
Direct Connect Semi-prep guard column holder(10mm) × 1, for 10×10mm Semi-Prep Cartridge