Learn More
Cayman Chemical SemiQuantitative Biomarker EL
SDP
Catalog No. 502886725
Encompass_Preferred
This assay contains a set of reagents to be used together with a suitable Biosense primary antibody for semi-quantitative detection of biomarkers such as Vtg, CYP1A, Zrp, and MT in samples from fish.
Catalog No. 50-288-6725 Supplier Cayman Chemical Supplier No. 100086591 EA
Add to Cart
Add to Cart
This assay contains a set of reagents to be used together with a suitable Biosense primary antibody for semi-quantitative detection of biomarkers such as Vtg, CYP1A, Zrp, and MT in samples from fish.