Learn More
USA Scientific Inc Semi-skt .1mL 96wl PCR plt
Catalog No. 502744870
Encompass
TempPlate semskrted polypropylene 0.1 mL 96-well PCR plate, low profile, straight sides, white. Two/sleeve, 10/box.
Catalog No. 50-274-4870 Supplier USA Scientific Inc Supplier No. 14029990
Add to Cart
Add to Cart
TempPlate semskrted polypropylene 0.1 mL 96-well PCR plate, low profile, straight sides, white. Two/sleeve, 10/box.