Learn More
Med Vet International Miltex Vantage Semken Dressing Forceps, 5in
Catalog No. 502559462
Encompass_Preferred
The Miltex Vantage V96-104 Semken Dressing Forceps are made of mid-grade stainless steel and have a length of 5in. Semken Dressing Forceps are commonly used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. This surgical forcep has straight blades with serrated handles.
Catalog No. 50-255-9462 Supplier Med Vet International Supplier No. MILV96104
The Miltex Vantage V96-104 Semken Dressing Forceps are made of mid-grade stainless steel and have a length of 5in. Semken Dressing Forceps are commonly used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. This surgical forcep has straight blades with serrated handles.