Learn More
Roboz Surgical SEMKEN FCP 5IN SERR
SDP
Catalog No. NC9193028
Encompass
Semken Forceps; Serrated; 1.2mm Tip Width; 5 in. Length Tissue forceps with fine serrations and stabilizing pin. Excellent forceps if an Adson-size tip is needed with greater length.
Catalog No. NC9193028 Supplier Roboz Surgical Supplier No. RS5240
Semken Forceps; Serrated; 1.2mm Tip Width; 5 in. Length Tissue forceps with fine serrations and stabilizing pin. Excellent forceps if an Adson-size tip is needed with greater length.